Test Strips, Zinc
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of Zinc in solutions. They come with all necessary reagents so that the measurement can be started immediately. A reliable result is obtained within 2 minutes.
Documents
Specifikationer
Specifikationer
- Pakn:100
- Beskrivelse:AQUADUR® 4-21
- Dimensioner:40×40×126 mm (packaging)
- Test pr. sæt:100 Tests
- Område:< 54 • >72 • >126 • >252 • >378 - ppm - CaCO₃; <3 • >4 • >7 • >14 • >21 - °e
- Indhold:100 test strips in a tube
- Cat. No.:662-0239
- Overensstemmelse:No hazardous material
- Platform:Semi-quantitative test strips
- Holdbarhed (måneder):24 (from production)
- Vægt:40 g
- Instrument:No
- Parameter:All reagents included
- Method:Total hardness - green → red
Specifikationer
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



