Orders LinkContactUsLinkComponent
Glass wool
Glass wool
Qualitative filter flocs, MN 101
Catalog # MANA481100
Supplier:   MACHEREY-NAGEL
Glass wool
Catalog # MANA481100
Supplier:  MACHEREY-NAGEL
Glass wool
Glass wool
Qualitative filter flocs, MN 101
Price
251,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Filter aids transform difficult precipitates and colloidal particles into a form which can be easily filtered. When slimy and strongly lyophilic, swelling precipitates are involved, the fibres of the filter flocs prevent formation of a continuous, impermeable layer on the filter. The resulting filter cake remains porous and permeable, and clogging of the filter is prevented.
Specifications
  • Beskrivelse:
    Qualitative filter flocs, MN 101
  • Seq:
    01