Orders LinkContactUsLinkComponent
Upright microscope, F1115
Upright microscope, F1115
Achromatic objective (spring), 60×/0,85
Catalog # 632-9061
Supplier:   MOTIC
Upright microscope, F1115
Catalog # 632-9061
Supplier:  MOTIC
Upright microscope, F1115
Upright microscope, F1115
Achromatic objective (spring), 60×/0,85
Price
428,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
F1115 er designet til applikationer inden for uddannelse.
  • High quality optics with achromatic objectives
  • Robust design for students
  • Efficient LED energy saving lamp
Delivery information: Leveres med støvdækken.
Specifications
  • Forstørrelse:
    60×/0,85
  • Beskrivelse:
    Achromatic objective (spring)
  • Seq:
    102
  • Cat. No.:
    632-9061
  • Revolver:
    4 positioner
  • Belysning:
    LED
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 632-9034
    Plastic carrying case
    Log in for availability
    1 * 1 Items - 460,00 kr