Upright microscope, F1115
Achromatic objective, 20×/0,40
Catalog # 632-9059
Supplier: MOTIC
Gå til:
Product Details & Documents
F1115 er designet til applikationer inden for uddannelse.
- High quality optics with achromatic objectives
- Robust design for students
- Efficient LED energy saving lamp
Delivery information: Leveres med støvdækken.
Documents
Specifikationer
- Forstørrelse:20×/0,40
- Beskrivelse:Achromatic objective
- Seq:101
- Cat. No.:632-9059
- Revolver:4 positioner
- Belysning:LED
Accessories
Produktinformation
- Catalog #BeskrivelseAvailabilityPrice