Orders LinkContactUsLinkComponent
Upright microscopes, 18 series
Upright microscopes, 18 series
Achromatic objective, 100×/1,25 (I)
Catalog # 632-8953
Supplier:   MOTIC
Upright microscopes, 18 series
Catalog # 632-8953
Supplier:  MOTIC
Upright microscopes, 18 series
Upright microscopes, 18 series
Achromatic objective, 100×/1,25 (I)
Price
444,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These upright microscopes are designed for use in education.
  • Efficient LED energy saving lamp
Ordering information: Supplied with dust cover and rechargeable batteries for 50 hours of use.
Specifications
  • Forstørrelse:
    100×/1,25 (I)
  • Beskrivelse:
    Achromatic objective
  • Seq:
    206
  • Cat. No.:
    632-8953
  • Belysning:
    LED
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Forstørrelse
    Availability
    Price
  • 632-9055
    Widefield eyepiece
    15× (12 mm)
    Log in for availability
    1 * 1 Items - 234,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Forstørrelse
    Availability
    Price
  • 632-9053
    Huygens eyepiece
    5×/14,5
    Log in for availability
    1 * 1 Items - 234,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Forstørrelse
    Availability
    Price
  • 632-8945
    Eyepiece
    10×/18
    Log in for availability
    1 * 1 Items - 234,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Forstørrelse
    Availability
    Price
  • 630-0081
    Widefield eyepiece
    20×/11
    Log in for availability
    1 * 1 Items - 428,00 kr