Orders LinkContactUsLinkComponent
Filter holders, Microsart®
Filter holders, Microsart®
Filter holder O-ring, silicone, 80×3 mm
Katalog # 514-1291
Leverandør:   Sartorius
Filter holders, Microsart®
Katalog # 514-1291
Leverandør:  Sartorius
Filter holders, Microsart®
Filter holder O-ring, silicone, 80×3 mm
Price
101,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
Spare parts for polycarbonate filter holder.
Spare parts for polycarbonate filter holder - individual filter holders, stand-alone, to be assembled on vacuum flasks or on manifolds, depending on the particular version.
 : Supplied without accessories.
Specifikationer
  • Beskrivelse:
    Filter holder O-ring, silicone, 80×3 mm
  • Seq:
    01