Orders LinkContactUsLinkComponent
Upright microscopes, 18 series
Mikrometer-okular, 100 inddelinger, 10 mm, WF 10×/18, 10×/18
  632-8946
 :   MOTIC
Upright microscopes, 18 series
  632-8946
 :  MOTIC
Upright microscopes, 18 series
Upright microscopes, 18 series
Mikrometer-okular, 100 inddelinger, 10 mm, WF 10×/18, 10×/18
524,00 kr
1 * 1 Items
These upright microscopes are designed for use in education.
  • Efficient LED energy saving lamp
 : Supplied with dust cover and rechargeable batteries for 50 hours of use.
  • Forstørrelse:
    10×/18
  • Beskrivelse:
    Mikrometer-okular, 100 inddelinger, 10 mm, WF 10×/18
  • Seq:
    103
  • Cat. No.:
    632-8946
  • Belysning:
    LED
Accessories
  •  
    Beskrivelse
    Forstørrelse
  • 632-9055
    Widefield eyepiece
    15× (12 mm)
    1 * 1 Items - 234,00 kr
  •  
    Beskrivelse
    Forstørrelse
  • 632-9053
    Huygens eyepiece
    5×/14,5
    1 * 1 Items - 234,00 kr
  •  
    Beskrivelse
    Forstørrelse
  • 632-8945
    Eyepiece
    10×/18
    1 * 1 Items - 234,00 kr
  •  
    Beskrivelse
    Forstørrelse
  • 630-0081
    Widefield eyepiece
    20×/11
    1 * 1 Items - 428,00 kr