Cuvettes for macrophotometers
45 mm, 2 ml, 10 mm, PS
Polystyrene cuvettes specially designed for macrophotometers.
- Længde på lysvej:10 mm
- Volumen:2 ml
- Vinduesmateriale:PS
- Højde:45 mm
- Seq:001
Polystyrene cuvettes specially designed for macrophotometers.