Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cuvettes for macrophotometers

Cuvettes for macrophotometers

45 mm, 4 ml, 10 mm, PS
  LPIT111117
Cuvettes for macrophotometers
  LPIT111117
Cuvettes for macrophotometers
Cuvettes for macrophotometers

45 mm, 4 ml, 10 mm, PS

 
1.110,00 kr
   
1 * 1.000 Items
 

Polystyrene cuvettes specially designed for macrophotometers.

 
  • Længde på lysvej:
    10 mm
  • Volumen:
    4 ml
  • Vinduesmateriale:
    PS
  • Højde:
    45 mm
  • Seq:
    002
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.