Membrane filters with grid
Hvid, med sort gitter, 135, 0,45 µm, 47 mm
Leverandør: Sartorius
Gå til:
Product Details & Documents
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Ø:47 mm
- Porestørrelse:0,45 µm
- Model:135
- Beskrivelse:Hvid, med sort gitter
- Seq:05
- Tykkelse:120 µm
- Cat. No.:515-0173
- Max. temperature:180 °C