Orders LinkContactUsLinkComponent
Membrane filters with grid
White with black grid, cellulose nitrate, 3 mm hydrophobic edge, 131, 0,45 µm, 50 mm
  SART13106-50-ACN
 :   Sartorius
Membrane filters with grid
  SART13106-50-ACN
 :  Sartorius
Membrane filters with grid
Membrane filters with grid
White with black grid, cellulose nitrate, 3 mm hydrophobic edge, 131, 0,45 µm, 50 mm
3.840,00 kr
1 * 100 Items
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.
  • Ø:
    50 mm
  • Porestørrelse:
    0,45 µm
  • Model:
    131
  • Beskrivelse:
    White with black grid, cellulose nitrate, 3 mm hydrophobic edge
  • Seq:
    07
  • Tykkelse:
    115 - 145 µm
  • Cat. No.:
    SART13106-50-ACN
  • Max. temperature:
    130 °C