Orders LinkContactUsLinkComponent
Membrane filters with grid
Membrane filters with grid
Hvid, med sort gitter, 131, 0,45 µm, 47 mm
Katalog # 611-0683
Leverandør:   Sartorius
Membrane filters with grid
Katalog # 611-0683
Leverandør:  Sartorius
Membrane filters with grid
Membrane filters with grid
Hvid, med sort gitter, 131, 0,45 µm, 47 mm
Price
2.050,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.
Documents
Specifikationer
  • Ø:
    47 mm
  • Porestørrelse:
    0,45 µm
  • Model:
    131
  • Beskrivelse:
    Hvid, med sort gitter
  • Seq:
    04
  • Tykkelse:
    115 - 145 µm
  • Cat. No.:
    611-0683
  • Max. temperature:
    130 °C