Orders LinkContactUsLinkComponent
Filterrør, Allihn
20 mm, 2, 30 ml, 9 mm
  LENZ07220002
Filterrør, Allihn
  LENZ07220002
Filterrør, Allihn
Filterrør, Allihn
20 mm, 2, 30 ml, 9 mm
233,00 kr
1 * 1 Items
Made of DURAN®, borosilicate glass 3,3.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
With integrated sintered glass filter plate.
  • Plade Ø:
    20 mm
  • Porøsitet:
    2
  • Kapacitet:
    30 ml
  • Stilk udv. Ø:
    9 mm