Orders LinkContactUsLinkComponent
Gas sampling tubes
Gas sampling tubes
500 ml, Uden udluftningstilslutning med gevind
Katalog # 201-9113
Leverandør:   Lenz Laborglas
Gas sampling tubes
Katalog # 201-9113
Leverandør:  Lenz Laborglas
Gas sampling tubes
Gas sampling tubes
500 ml, Uden udluftningstilslutning med gevind
Price
959,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Duran® clear borosilicate glass.
  • Excellent thermal and chemical resistance
  • Connection on one side with capillary tube (internal Ø: 2,7 mm, external Ø: 8,0 mm) and on the other side with tube connector (external Ø: 8,0 mm)
  • With white labelling area
Gas collection tubes for removing, collecting and transporting gas samples. Both sides with a 3 NS/14 standard ground one-way stop cock.
Certifications: DIN 12473-1
Documents
Specifikationer
  • Model:
    Uden udluftningstilslutning med gevind
  • Kapacitet:
    500 ml