Tørrerør med standardslib, vinklede
14/23 NS
Katalog # 201-5311
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
Clear DURAN® borosilicate 3.3 glass. Angled at 75° with ground joint cone.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Konusstr.:14/23 NS