Orders LinkContactUsLinkComponent
Distilling receivers, Bernauer
Distilling receivers, Bernauer
14/23 NS
Katalog # 201-0906
Leverandør:   Lenz Laborglas
Distilling receivers, Bernauer
Katalog # 201-0906
Leverandør:  Lenz Laborglas
Distilling receivers, Bernauer
Distilling receivers, Bernauer
14/23 NS
Price
790,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Documents
Specifikationer
  • Kappestr.:
    14/23 NS