Orders LinkContactUsLinkComponent
Adapters, delivery, bent
Adapters, delivery, bent
29/32 NS, 29/32 NS
Katalog # 201-3110
Leverandør:   Lenz Laborglas
Adapters, delivery, bent
Katalog # 201-3110
Leverandør:  Lenz Laborglas
Adapters, delivery, bent
Adapters, delivery, bent
29/32 NS, 29/32 NS
Price
235,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Documents
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS