Orders LinkContactUsLinkComponent
Adapters, delivery, bent
19/26 NS, 19/26 NS
Catalog # 201-3109
Supplier:   Lenz Laborglas
Adapters, delivery, bent
Catalog # 201-3109
Supplier:  Lenz Laborglas
Adapters, delivery, bent
Adapters, delivery, bent
19/26 NS, 19/26 NS
Price
192,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Specifications
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    19/26 NS