Orders LinkContactUsLinkComponent
Splash heads, Stutzer, bent
Splash heads, Stutzer, bent
29/32 NS
Katalog # 271-2231
Leverandør:   Lenz Laborglas
Splash heads, Stutzer, bent
Katalog # 271-2231
Leverandør:  Lenz Laborglas
Splash heads, Stutzer, bent
Splash heads, Stutzer, bent
29/32 NS
Price
798,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.
Documents
Specifikationer
  • Konusstr.:
    29/32 NS