Splash heads, Stutzer, bent
19/26 NS
201-0343
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
- Konusstr.:19/26 NS