Dråbefangere, Stutzer, lige
29/32 NS, 29/32 NS
Katalog # 201-2232
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Anbringes direkte på destillationskolben for at fange dråber, med to standardslib.
Specifikationer
- Konusstr.:29/32 NS
- Kappestr.:29/32 NS
- Seq:04