Dråbefangere, Stutzer, lige
19/26 NS, 24/29 NS
Katalog # 201-0342
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Anbringes direkte på destillationskolben for at fange dråber, med to standardslib.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Konusstr.:24/29 NS
- Kappestr.:19/26 NS
- Seq:03