Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
24/29 NS, 24/29 NS
Katalog # 201-0337
Leverandør:   Lenz Laborglas
Multiple adapters
Katalog # 201-0337
Leverandør:  Lenz Laborglas
Multiple adapters
Multiple adapters
24/29 NS, 24/29 NS
Price
371,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two sockets and one cone, standard ground joints and two parallel necks.
Documents
Specifikationer
  • Konusstr.:
    24/29 NS
  • Kappestr.:
    24/29 NS