Orders LinkContactUsLinkComponent
Skilletragte, Gilson
250 ml, 29/32 NS
Catalog # 527-1503
Supplier:   Lenz Laborglas
Skilletragte, Gilson
Catalog # 527-1503
Supplier:  Lenz Laborglas
Skilletragte, Gilson
Skilletragte, Gilson
250 ml, 29/32 NS
Price
759,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilikatglas, med NS-PE prop.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Specifications
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    250 ml