Skilletragte, Dråbeformet
2000 ml, 29/32 NS, -
Katalog # 527-1326
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilikatglas, med NS-PE prop.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.
Specifikationer
- Kappestr.:29/32 NS
- Kapacitet:2000 ml
- Inddeling:-