Koniske skilletragte
1000 ml, 29/32 NS, -
Katalog # 527-1140
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilikatglas, med NS-PE prop.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
LENZ04000073_TDS_EN
Datablad
Specifikationer
- Kappestr.:29/32 NS
- Kapacitet:1000 ml
- Inddeling:-
- Cat. No.:527-1140