Gel loading-spidser, ART™
ART 100 GEL, 100 µl, +, +, 68,6 mm, 10 racks, 96 pr. stk.
Leverandør: Thermo Fisher Scientific
Gel loading-spidser, ART™
Katalog # 732-2226
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
These tips are designed for loading vertical acrylamide or agarose gels.
- Sterile tips are certified free from RNase, DNase, ATP and pyrogen
- Available with or without filter for trapping potential contaminants
- Manufactured according to the industry's highest quality standard
Documents
Specifikationer
- Pakket:10 racks, 96 pr. stk.
- Steril:+
- Volumen:100 µl
- Længde:68,6 mm
- Seq:03
- Spidstype:ART 100 GEL
- Filter:+
- Cat. No.:732-2226
- Miljøcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lav fremstillingspåvirkning