Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pressure vessels

Pressure vessels

20 l, Dispensing pressure vessel, for holding liquid for filtration
Katalog # MILFXX6700P20
Leverandør:   Merck
Pressure vessels
Katalog # MILFXX6700P20
Leverandør:  Merck
Pressure vessels
Pressure vessels

20 l, Dispensing pressure vessel, for holding liquid for filtration

Price
17.550,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Dispensing pressure vessels hold liquid for filtration through pressure-operated filter holders, if connected to an external pressure source. Used for large-volume filtration, reservoir for buffer or solvent dispensing.

  • Made from stainless steel 316
  • Closures are secured by cam-lock handle
  • Vessels are autoclavable when vented
  • Fluoroelastomer gaskets and O-rings used
  • Molded neoprene rubber base

Max. operating pressure: 6,9 bar

Max. operating temperature: 121 °C

Certifications: Meets ASME®-UM Code requirements.

Specifikationer
  • Kapacitet:
    20 l
  • Beskrivelse:
    Dispensing pressure vessel, for holding liquid for filtration
  • Seq:
    04
Accessories

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • O-ring, fluoroelastomer A
    O-ring, fluoroelastomer A
    MILFXX6700059
    Log ind for tilgængelighed
    1 * 1 Items - kr1,320.00

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Bubble point test apparatus
    Bubble point test apparatus
    MILFXX6700L55
    Log ind for tilgængelighed
    1 * 1 Items - kr12,590.00

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Pressure tubing, PVC, I.Ø 12,7 mm
    Pressure tubing, PVC, I.Ø 12,7 mm
    MILFXX6700035
    Log ind for tilgængelighed
    1 * 1 Items - kr1,020.00

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Pressure vessel accessory kit
    Pressure vessel accessory kit
    MILFXX67000PK
    Log ind for tilgængelighed
    1 * 1 Items - kr14,290.00

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Plug, stainless steel, 6,35 mm NPT
    Plug, stainless steel, 6,35 mm NPT
    MILFYY1301009
    Log ind for tilgængelighed
    1 * 1 Items - kr448.91

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Quick-release nipple and coupling, 6,35 mm NPTM
    Quick-release nipple and coupling, 6,35 mm NPTM
    MILFXX6700030
    Log ind for tilgængelighed
    1 * 1 Items - kr1,498.49

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Pressure gauge, 6,35 mm ASME
    Pressure gauge, 6,35 mm ASME
    MILFXX6700L15
    Log ind for tilgængelighed
    1 * 1 Items - kr1,970.00

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Vent/relief valve, 7 bar, ASME
    Vent/relief valve, 7 bar, ASME
    MILFXX6700L24
    Log ind for tilgængelighed
    1 * 1 Items - kr2,260.00

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Adapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.Ø
    Adapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.Ø
    MILFXX6700L11
    Log ind for tilgængelighed
    1 * 2 Items - kr2,280.00