Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Stirrer blades for balloon flasks

Stirrer blade, PTFE, for 12 - 22 l balloon flask, 149,05 mm
Katalog # KART694
Leverandør:   KARTELL
Stirrer blades for balloon flasks
Katalog # KART694
Leverandør:  KARTELL
Stirrer blades for balloon flasks
Stirrer blades for balloon flasks

Stirrer blade, PTFE, for 12 - 22 l balloon flask, 149,05 mm

Price
185,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Fit all standard button tip stirring shafts. 'Long groove' keyhole (Ø 9 mm) provides easy insertion into flask neck and proper position for stirring action. Can be used continuously at temperatures up to 250 °C and, for short intervals, to 300 °C.

Specifikationer
  • Model:
    Stirrer blade, PTFE, for 12 - 22 l balloon flask
  • Bredde:
    149,05 mm