Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Stirrer blades for balloon flasks

Stirrer blades for balloon flasks

Stirrer blade, PTFE, for 5 l balloon flask, 122,53 mm
Katalog # KART693
Leverandør:   KARTELL
Stirrer blades for balloon flasks
Katalog # KART693
Leverandør:  KARTELL
Stirrer blades for balloon flasks
Stirrer blades for balloon flasks

Stirrer blade, PTFE, for 5 l balloon flask, 122,53 mm

Price
165,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Fit all standard button tip stirring shafts. 'Long groove' keyhole (Ø 9 mm) provides easy insertion into flask neck and proper position for stirring action. Can be used continuously at temperatures up to 250 °C and, for short intervals, to 300 °C.

Specifikationer
  • Model:
    Stirrer blade, PTFE, for 5 l balloon flask
  • Bredde:
    122,53 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.