Orders LinkContactUsLinkComponent
Stirrer blades for balloon flasks
Stirrer blade, PTFE, for 1 l balloon flask, 75, 23 mm
  KART691
 :   KARTELL
Stirrer blades for balloon flasks
  KART691
 :  KARTELL
Stirrer blades for balloon flasks
Stirrer blades for balloon flasks
Stirrer blade, PTFE, for 1 l balloon flask, 75, 23 mm
140,00 kr
1 * 1 Items
Fit all standard button tip stirring shafts. 'Long groove' keyhole (Ø 9 mm) provides easy insertion into flask neck and proper position for stirring action. Can be used continuously at temperatures up to 250 °C and, for short intervals, to 300 °C.
  • Model:
    Stirrer blade, PTFE, for 1 l balloon flask
  • Bredde:
    75, 23 mm