Orders LinkContactUsLinkComponent
Drying tubes, calcium chloride
8-10 mm, 270 mm, 20x200 mm
  468-1212
 :   KARTELL
Drying tubes, calcium chloride
  468-1212
 :  KARTELL
Drying tubes, calcium chloride
Drying tubes, calcium chloride
8-10 mm, 270 mm, 20x200 mm
kr38.70
1 * 1 Items
PE.
  • Both ends have tapered, push-on connectors
Suitable for drying gases.
  • Ø×L:
    20x200 mm
  • Til rør Ø:
    8-10 mm
  • Samlet længde:
    270 mm