Weighing bottles, PP
20 ml, 40×29 mm
Katalog # 611-9402
Leverandør: KARTELL
Gå til:
Product Details & Documents
Fremstillet i PP med låg.
- Very good chemical resistance
- Very good temperature resistance
Cylindrical shape, with knobbed lid.
Documents
Specifikationer
- Diameter×H:40×29 mm
- Kapacitet:20 ml