Orders LinkContactUsLinkComponent
Polyethylene threaded water bottle caps
250/500 ml, 215-1331/215-1332, GL 25
Katalog # 217-0486
Leverandør:   KARTELL
Polyethylene threaded water bottle caps
Katalog # 217-0486
Leverandør:  KARTELL
Polyethylene threaded water bottle caps
Polyethylene threaded water bottle caps
250/500 ml, 215-1331/215-1332, GL 25
Price
19,70 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
LDPE.
  • Fine tip tube
Specifikationer
  • Til:
    215-1331/215-1332
  • Gevind:
    GL 25
  • Seq:
    02
  • Til flasker:
    250/500 ml