Bottle top tilt measures
25 /500 ml, gennemsigtig
Katalog # KART1476
Leverandør: KARTELL
Gå til:
Product Details & Documents
PE (bottle and tube), PMP (measuring cup).
- Repetitive dispensing of the same volume of liquid
- When the bottle pressure is released only the desired volume remains
The 25 /50 ml measuring cup slides up and down the tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
Colour: Transparent /Red
Specifikationer
- Kapacitet:25 /500 ml
- Farve:gennemsigtig