Orders LinkContactUsLinkComponent
Bottle top tilt measures
Bottle top tilt measures
25 /500 ml, gennemsigtig
Katalog # KART1476
Leverandør:   KARTELL
Bottle top tilt measures
Katalog # KART1476
Leverandør:  KARTELL
Bottle top tilt measures
Bottle top tilt measures
25 /500 ml, gennemsigtig
Price
87,90 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PE (bottle and tube), PMP (measuring cup).
  • Repetitive dispensing of the same volume of liquid
  • When the bottle pressure is released only the desired volume remains
The 25 /50 ml measuring cup slides up and down the tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
Colour: Transparent /Red
Specifikationer
  • Kapacitet:
    25 /500 ml
  • Farve:
    gennemsigtig