Orders LinkContactUsLinkComponent
Draining racks, with drip pan
Draining racks, with drip pan
Draining rack, with 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm), 420×160×610 mm
Katalog # BOCH10140
Leverandør:   BOCHEM
Draining racks, with drip pan
Katalog # BOCH10140
Leverandør:  BOCHEM
Draining racks, with drip pan
Draining racks, with drip pan
Draining rack, with 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm), 420×160×610 mm
Price
1.670,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These PA coated wire draining racks are suitable for test tubes and flasks.
  • With drip pan
  • Good chemical resistance
  • Good temperature resistance
Specifikationer
  • Beskrivelse:
    Draining rack, with 24 rods (95 mm) and 20 arches (2×110 mm and 3×70 mm)
  • Seq:
    01
  • B×D×H:
    420×160×610 mm