Drug test kit
Cannabis reagens 1
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
- Kittet består af følgende:
- 20 (4 boxes of 5, 0.2 ml) ampoules of marquis reagent
- 1×10 ml kokain reagens
- 1×10 ml barbiturat reagens
- 1×10 ml LSD reagens
- 1×20 ml cannabis reagens 1
- 1×10 ml cannabis reagens 2
- 1×5 g cannabis special solid reagens
- 1x20 g neutralisator
- 4 boxes of Whatman filterpapir
- 1 mikro-spatel
- 20 ampul breakers
- 25 engangsspatler
- pincetter
- 4 urglas
- Instruktionsmanual
Specifikationer
- Beskrivelse:Cannabis reagens 1
- Seq:03