Glycin 99+% for analysis
Plastic bottle, 250 g
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :13,04504
- :181 °C (1013 hPa)
- :240 °C
- :1,595 g/cm³
- :Stue temperatur
- :200-272-2
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 2 h)
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥99.0 % (on dried substance)
- Chloride (Cl):≤40 ppm
- Sulfate (SO4):≤50 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Hydrolyzable substance:Clear solution
- Residual solvents:Meets the requirements
- Pakning:Plastic bottle
- Pakn:250 g



