Glycin 99+% for analysis
Plastic bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Glycin 99+% for analysis
Katalog # ACRO220910010
Leverandør: Thermo Fisher Scientific
CAS nummer: 56-40-6
Gå til:
Product Details & Documents
Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS nummer:56-40-6
- Formel:NH₂CH₂COOH
- MDL nummer:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Kogende Pt:181 °C (1013 hPa)
- Smeltende Pt:240 °C
- Tæthed:1,595 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-272-2
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 2 h)
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥99.0 % (on dried substance)
- Chloride (Cl):≤40 ppm
- Sulfate (SO4):≤50 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Hydrolyzable substance:Clear solution
- Residual solvents:Meets the requirements
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



