(±)-2-Butanol 99+% for analysis
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
(±)-2-Butanol 99+% for analysis
Katalog # ACRO220290010
Leverandør: Thermo Fisher Scientific
CAS nummer: 78-92-2
Gå til:
Product Details & Documents
Synonymer: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
- CAS nummer:78-92-2
- Formel:H₃CCH₂CH(OH)CH₃
- MDL nummer:MFCD00004569
- Molecular Weight:74.12 g/mol
- Merck Index:13,01540
- Kogende Pt:99 °C (760 mmHg)
- Smeltende Pt:–115 °C
- Tæthed:0,81 g/cm³
- Flash Pt:24 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-158-5
- FN:1120
- ADR:3,III
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.2 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Non-volatile matter:≤0.001 %
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.1 ppm
- Free acid (C4H8O2):≤0.003 %
- 2-Propanol:≤0.2 % (GC)
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L



