Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-thyroxine 97+%

L-thyroxine 97+%

Glass bottle, 5 g
Katalog # ACRO215660050
Leverandør:   Thermo Fisher Scientific
L-thyroxine 97+%
Katalog # ACRO215660050
CAS nummer:  51-48-9
L-thyroxine 97+%
L-thyroxine 97+%

Glass bottle, 5 g

CAS nummer:  51-48-9
Price
5.000,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS nummer:51-48-9
  • Formel:C₁₅H₁₁I₄NO₄
  • MDL nummer:MFCD00002595
  • Molecular Weight:776.874 g/mol
  • Smeltende Pt:235…236 °C (dec.)
  • Opbevaringstemperatur:Køleskab

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1.0 % (110°C, 4 h)
  • HPLC:
    ≥97.0 %
  • Specific optical rotation:
    -4.0° to -6.0° (20°C, 589 nm) (c=1,1N NaOH:EtOH 1:2) (on dried substance)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.