Linolsyre 99%
Glass bottle, 25 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- CAS nummer:60-33-3
- Formel:C₁₈H₃₂O₂
- MDL nummer:MFCD00064241
- Molecular Weight:280.45 g/mol
- Merck Index:13,05527
- Kogende Pt:229…230 °C (16 mmHg)
- Smeltende Pt:–5 °C
- Tæthed:0,9 g/cm³
- Flash Pt:>112 °C
- Opbevaringstemperatur:Køleskab
- EINECS:200-470-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4687 to 1.4707 (20°C, 589 nm)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:25 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



