Linolsyre 99%
Glass bottle, 5 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- CAS nummer:60-33-3
- Formel:C₁₈H₃₂O₂
- MDL nummer:MFCD00064241
- Molecular Weight:280.45 g/mol
- Merck Index:13,05527
- Kogende Pt:229…230 °C (16 mmHg)
- Smeltende Pt:–5 °C
- Tæthed:0,9 g/cm³
- Flash Pt:>112 °C
- Opbevaringstemperatur:Køleskab
- EINECS:200-470-9
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4687 to 1.4707 (20°C, 589 nm)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL



