Kaliumfluorid 99+% for analysis
Plastic bottle, 250 g
Leverandør: Thermo Fisher Scientific
Kaliumfluorid 99+% for analysis
Katalog # ACRO212602500
Leverandør: Thermo Fisher Scientific
CAS nummer: 7789-23-3
Gå til:
Product Details & Documents
- CAS nummer:7789-23-3
- Formel:KF
- MDL nummer:MFCD00011398
- Molecular Weight:58.1 g/mol
- Merck Index:13,07714
- Kogende Pt:1505 °C (1013 hPa)
- Smeltende Pt:860 °C
- Tæthed:2,48 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-151-5
- FN:1812
- ADR:6.1,III
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Titration:≥99.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤200 ppm
- Potassium fluosilicate:≤0.1 %
- Cadmium (Cd):≤5 ppm
- Cobalt (Co):≤5 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤5 ppm
- Lead (Pb):≤5 ppm
- Nickel (Ni):≤5 ppm
- Zinc (Zn):≤5 ppm
- Free acid (HF):≤0.01 %
- Free alkali:≤0.01 % (K2CO3)
Specifikationer
- Pakning:Plastic bottle
- Pakn:250 g



