Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Basic Violet 3, pure indicator

Glass bottle for solids, 25 g
Katalog # ACRO212120250
Irriterende
Sundhed
Ætsende
Miljø
Leverandør:   Thermo Fisher Scientific
Basic Violet 3, pure indicator
Katalog # ACRO212120250
CAS nummer:  548-62-9
Basic Violet 3, pure indicator
Basic Violet 3, pure indicator

Glass bottle for solids, 25 g

CAS nummer:  548-62-9
Price
308,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
  • CAS nummer:548-62-9
  • Formel:C₂₅H₃₀ClN₃
  • MDL nummer:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Smeltende Pt:173 °C (dec.)
  • Tæthed:1,19 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-953-6
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark green Crystalline powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Sensitivity:
    (as indicator) Passes test

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer