Test sets
Test set I for inorganic gases
Katalog # 151-0176
Leverandør: Dräger
Gå til:
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifikationer
- Beskrivelse:Test set I for inorganic gases
- Seq:01
- Cat. No.:151-0176
- Model:Test set I for inorganic gases
- Måletid:40 s