Orders LinkContactUsLinkComponent
Test sets
Test sets
Adapter for simultaneous test via connection to a bellows pump
Katalog # 151-0116
Leverandør:   Dräger
Test sets
Katalog # 151-0116
Leverandør:  Dräger
Test sets
Test sets
Adapter for simultaneous test via connection to a bellows pump
Price
680,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifikationer
  • Beskrivelse:
    Adapter for simultaneous test via connection to a bellows pump
  • Seq:
    07