Procain, HCl 99%
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride
- CAS nummer:51-05-8
- Formel:C₁₃H₂₁ClN₂O₂
- MDL nummer:MFCD00013000
- Molecular Weight:272.77 g/mol
- Smeltende Pt:154…158 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-077-2
- FN:2811
- ADR:6.1,III
Specifikationer
Specifikationstestresultater
- Appearance:Colorless to white to almost white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:154 to 158 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- Sulfated ash:≤0.1 %
- pH:5.0 to 6.5 (2 % w/v in water)
- Titration with NaOH:98.5 to 101.5 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g



