Bariumnitrat 99+% for analysis
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Bariumnitrat 99+% for analysis
Katalog # ACRO203155000
Leverandør: Thermo Fisher Scientific
CAS nummer: 10022-31-8
Gå til:
Product Details & Documents
- CAS nummer:10022-31-8
- Formel:Ba(NO₃)₂
- MDL nummer:MFCD00003442
- Molecular Weight:261.34 g/mol
- Merck Index:13,00986
- Smeltende Pt:592 °C
- Tæthed:~ 3,23 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:233-020-5
- FN:1446
- ADR:5.1,II
Documents
ACRO20315
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Crystals
- Titration Complexometric:≥99.0 %
- pH:5 to 8 (5 % at 20°C)
- Chloride (Cl):≤10 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤5 ppm
- Strontium (Sr):≤1000 ppm
- Heavy metals (as Pb):≤10 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



