Tannin 95%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Gallotannin, Tannin
- CAS nummer:1401-55-4
- Formel:C₇₆H₅₂O₄₆
- MDL nummer:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Smeltende Pt:218 °C (dec.)
- Tæthed:Min, 1 g/cm³ (20 °C)
- Flash Pt:198 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-753-2
Specifikationer
Specifikationstestresultater
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Assay:≥94 % (on dry substance)
- Loss on drying:≤12.0 %
- Sulfated ash:≤1.0 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



